Polyethylene glycol 8,000

Supplier: Roth Carl

ROTH0263.1EA 54.1 EUR
ROTH0263.1 ROTH0263.2 ROTH0263.3
Polyethylene glycol 8,000
Polyethylene glycol 8,000
Formula: H(OCH₂CH₂)nOH
MW: 8000 g/mol
Boiling Pt: 250 °C (1013 hPa)
Density: 1,125 g/cm³ (20 °C)
MDL Number: MFCD01779615
CAS Number: 25322-68-3
Merck Index: 12,07729

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR