Polyethylene glycol 8,000, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ200
81268-1KG
81268-250G
81268-5KG
SIAL81268-1KGEA
157
EUR
SIAL81268-1KG
SIAL81268-250G
SIAL81268-5KG
Polyethylene glycol 8,000, Sigma-Aldrich®
Polyethylene glycol 8,000
Formula:
H(OCH₂CH₂)nOH Boiling Pt: 250 °C (1013 hPa) Density: 1.125 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD01779615 CAS Number: 25322-68-3 EINECS: 500-038-2 Merck Index: 12,07729 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...