TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)
Supplier: G Bioscienes
DG007EA 280 EUR
DG007
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)
TRITON™ X-100 (Polyethylene glycol tert-octylphenyl ether)
Boiling Pt: ∼200 °C (1013 hPa)
Melting Pt: 6 °C
Density: 1,06 g/cm³ (20 °C)
Flash Pt: 251 °C (closed cup)
MDL Number: MFCD00132505
UN: 3082
ADR: 9,III
Merck Index: 13,06793