TRITON™ X-405 (Polyethylene glycol tert-octylphenyl ether), Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ100
X405-100ML X405-1L X405-500ML X405-1GA X405-5GA
SIALX405-100MLEA 66.3 EUR
SIALX405-100ML SIALX405-1L SIALX405-500ML SIALX405-1GA SIALX405-5GA
TRITON™ X-405 (Polyethylene glycol tert-octylphenyl ether), Sigma-Aldrich®
TRITON™ X-405 (Polyethylene glycol tert-octylphenyl ether)
Formula: C₂₈H₅₀O₈
Boiling Pt: 120 °C (1013 hPa)
Melting Pt: –4 °C
Density: 1.05 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD01779734
CAS Number: 9002-93-1
Merck Index: 12,06858