Paraformaldehyde, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Polyoxymethylene
M-Clarity™ quality level = MQ200
441244-1KG 441244-3KG
SIAL441244-1KGEA 44.4 EUR
SIAL441244-1KG SIAL441244-3KG
Paraformaldehyde, Sigma-Aldrich®
Paraformaldehyde
Formula: (CH₂O)n
Boiling Pt: 135…136 °C (13 mmHg)
Melting Pt: 123
Density: 1.46 g/cm³ (20 °C)
Flash Pt: 70
Storage Temperature: Ambient
MDL Number: MFCD00133991
CAS Number: 30525-89-4
UN: 2213
ADR: 4.1 III
Merck Index: 13,07096