Polysorbate 80
Supplier: Thermo Fisher Scientific
Synonyms: Sorbitan monooleate, ethoxylated, Polyoxyethylene-80-Sorbitan Monolaurate, Polyoxyethylene (20) sorbitan monooleate
L13315.AE L13315.AP L13315.0F
L13315.AEEA 33 EUR
L13315.AE L13315.AP L13315.0F
Polysorbate 80
Polysorbate 80
CAS No.: 9005-65-6 Flash Point: 149°(300°F) Refractive Index: 1.4750 Boiling Point: 100° Merck Reference: 14,7582 MDL No.: MFCD00082107
Boiling Pt: >100 °C
Melting Pt: 100
Density: 1.064
Flash Pt: >149 °C (300 °F)
Storage Temperature: Ambient
MDL Number: MFCD00082107
CAS Number: 9005-65-6
EINECS: 500-019-9
Merck Index: 14,7582