Polysorbate 80, Sigma-Aldrich®
Supplier: Merck
Synonyms: Sorbitan monooleate, ethoxylated, Polyoxyethylene-80-Sorbitan Monolaurate, Polyoxyethylene (20) sorbitan monooleate
MQ200
59924-100G-F 59924-1KG-F
SIAL59924-100G-FEA 31.7 EUR
SIAL59924-100G-F SIAL59924-1KG-F
Polysorbate 80, Sigma-Aldrich®
Polysorbate 80
Melting Pt: 100
Flash Pt: 113 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00082107
CAS Number: 9005-65-6
EINECS: 500-019-9