Tween® 20 (Polysorbate) Molecular_Biology
Supplier: PanReac AppliChem
Synonyms: Polyoxyethylene sorbitan monolaurate
A4974.0500 A4974.1000
A4974.0500EA 64.5 EUR
A4974.0500 A4974.1000
Tween® 20 (Polysorbate) Molecular_Biology
Tween® 20 (Polysorbate)
Formula: C₅₈H₁₁₄O₂₆
MW: 1227.53 g/mol
Density: 1.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00165986
CAS Number: 9005-64-5
EINECS: 500-018-3
Order Now
Specification Test Results
DNases/RNases/Proteases not detectable
Density (d 20°C/4°C) 1.095 - 1.105
Hydroxyl-No. approx. 100
Saponification No. 40 - 50