Tween® 20 (Polysorbate)
Supplier: Roth Carl
Synonyms: Polyoxyethylene sorbitan monolaurate
9127.1 9127.2
ROTH9127.1EA 29.5 EUR
ROTH9127.1 ROTH9127.2
Tween® 20 (Polysorbate)
Tween® 20 (Polysorbate)
Formula: C₅₈H₁₁₄O₂₆
MW: 1227.53 g/mol
Density: 1.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00165986
CAS Number: 9005-64-5
EINECS: 500-018-3