Polysorbate 20, Sigma-Aldrich®
Supplier: Merck
Synonyms: Polyoxyethylene-20-Sorbitan Monolaurate, Sorbitan monolaurate, ethoxylated
M-Clarity™ quality level = MQ300
P1379-100ML P1379-500ML P1379-1L P1379-25ML
SIALP1379-100MLEA 37.5 EUR
SIALP1379-100ML SIALP1379-500ML SIALP1379-1L SIALP1379-25ML
Polysorbate 20, Sigma-Aldrich®
Polysorbate 20
Formula: C₅₈H₁₁₄O₂₆
Storage Temperature: Ambient
MDL Number: MFCD00165986
CAS Number: 9005-64-5
EINECS: 500-018-3