Polyvinylpyrrolidone, M.W. ∼ 24,000 (K25)
Supplier: Roth Carl
Synonyms:
Povidone, Polyvinylpyrrolidon, Polyvidone, PVP
4606.1
4606.2
ROTH4606.1EA
28.9
EUR
ROTH4606.1
ROTH4606.2
Polyvinylpyrrolidone, M.W. ∼ 24,000 (K25)
Polyvinylpyrrolidone
Formula:
(C₆H₉NO)n Storage Temperature: Ambient |
MDL Number:
MFCD01076626 CAS Number: 9003-39-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...