Dichloromethane 99,8 Stabilised, PESTINORM® Pest_Residual
Supplier: VWR Chemicals
Warning
Synonyms: Methylene chloride
Stabilization: 20 ppm 2-Methyl-2-buten
Filter 0,2 um_Nitrogen_filled
83665.320EA 164 EUR
83665.320 83665.290 83665.400
Dichloromethane 99,8 Stabilised, PESTINORM® Pest_Residual
Dichloromethane
Formula: CH₂Cl₂
MW: 84.93 g/mol
Boiling Pt: 39.8 °C (1013 hPa)
Melting Pt: –95 °C
Density: 1.322 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00000881
CAS Number: 75-09-2
EINECS: 200-838-9
UN: 1593
ADR: 6.1 III
REACH: 01-2119480404-41
Merck Index: 12,06140
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=bea1f8abe149a925f126e308d8aaee3a107495c9&vid=1a50c1b5a88f4f946c811d278fe40566f022efb4&prid=web&clid=SAPDAM
Order Now
Specification Test Results
Assay (on anhydrous substance) Min. 99,8%
Water Max. 0,02 %
Evaporation residue Max. 0,0005 %
Pesticide analysis (Lindane/ECD) Max. 5 ng/l