(±)-2,3-Dibromo-1-propanol, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: 2,3-Dibromo-1-propanol, Glycerol alpha,beta-Dibromohydrin, alpha,beta-Dibromohydrin

M-Clarity™ quality level = MQ200

D43050-100G D43050-500G
SIALD43050-100GEA 39.9 EUR
SIALD43050-100G SIALD43050-500G
(±)-2,3-Dibromo-1-propanol, Sigma-Aldrich®
(±)-2,3-Dibromo-1-propanol
Formula: HOCH₂BrCHCH₂Br
MW: 217.89 g/mol
Boiling Pt: 219 °C (1013 hPa)
Melting Pt: 8 °C
Density: 2.1682 g/cm³ (0 °C)
MDL Number: MFCD00004699
CAS Number: 96-13-9
EINECS: 202-480-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR