(±)-3-Chloro-1,2-propanediol, Sigma-Aldrich®
Supplier: Merck
Synonyms: (±)-3-Chloro-1,2-dihydroxy propane
M-Clarity™ quality level = MQ100
107271-500ML 107271-25ML
SIAL107271-500MLEA 73.6 EUR
SIAL107271-500ML SIAL107271-25ML
(±)-3-Chloro-1,2-propanediol, Sigma-Aldrich®
(±)-3-Chloro-1,2-propanediol
Formula: C₃H₇ClO₂
Boiling Pt: 213 °C (1013 hPa)
Melting Pt: –40 °C
Density: 1.32 g/cm³ (20 °C)
Flash Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00004712
CAS Number: 96-24-2
EINECS: 202-492-4
UN: 2689
ADR: 6.1,III
Merck Index: 13,02164