1-Iodododecane ≥98% stabilised
Supplier: Thermo Scientific
Warning
Synonyms: 1-Dodecyl iodide
Stabilization: Stabilised with copper
A11909.14 A11909.22 A11909.36
A11909.14EA 49.9 EUR
A11909.14 A11909.22 A11909.36
1-Iodododecane ≥98% stabilised
1-Iodododecane
Stabilized with copper
Purity: 98% CAS No.: 4292-19-7 Molecular Formula: CH3(CH2)11I Formula Weight: 296.24 Flash Point: 110°(230°F) Refractive Index: 1.4844 Melting Point: -3° Boiling Point: 128-130°/11mm Beilstein Reference: 1738943 MDL No.: MFCD00001088
Formula: C₁₂H₂₅I
MW: 296.24 g/mol
Boiling Pt: 128…130 °C (11 mmHg)
Melting Pt: –3 °C
Density: 1.201
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001088
CAS Number: 4292-19-7
EINECS: 224-293-1