L(+)-Arginine ≥98.5%, crystalline powder Ph. Eur., USP Ajinomoto cell culture reagent, GMP, SAFC®
Supplier: Merck
Warning
M-Clarity™ quality level = MQ500 (more info.)
A4474-100G A4474-10KG A4474-25KG
SIALA4474-100GEA 0 EUR
SIALA4474-100G SIALA4474-10KG SIALA4474-25KG
L(+)-Arginine ≥98.5%, crystalline powder Ph. Eur., USP Ajinomoto cell culture reagent, GMP, SAFC®
L(+)-Arginine
Formula: C₆H₁₄N₄O₂
Boiling Pt: 368 °C (1013 hPa)
Melting Pt: 238 °C
Density: 1.3 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002635
CAS Number: 74-79-3
EINECS: 200-811-1
Merck Index: 13,00785