DL-Homocysteine, Sigma-Aldrich®
Supplier: Merck
Synonyms:
(±)-Homocysteine, 2-Amino-4-mercaptobutanoic acid, H-DL-HoCys-OH
M-Clarity™ quality level = MQ100
H4628-10MG
H4628-1G
H4628-25G
H4628-5G
SIALH4628-10MGEA
37.9
EUR
SIALH4628-10MG
SIALH4628-1G
SIALH4628-25G
SIALH4628-5G
DL-Homocysteine, Sigma-Aldrich®
DL-Homocysteine
Formula:
C₄H₉NO₂S MW: 135.19 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00004898 CAS Number: 454-29-5 EINECS: 207-222-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...