DL-Malic acid, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: DL-Hydroxysuccinic acid, (±)-Malic acid, (±)-Hydroxy-1,4-butanedioic acid, (±)-Hydroxysuccinic acid, Hydroxybutanedioic acid, 2-Hydroxysuccinic acid
M-Clarity™ quality level = MQ300
W265501-10KG-K W265501-1KG-K W265501-5KG-K
SIALW265501-10KG-KEA 165 EUR
SIALW265501-10KG-K SIALW265501-1KG-K SIALW265501-5KG-K
DL-Malic acid, Sigma-Aldrich®
DL-Malic acid
Formula: HOOCCH(OH)CH₂COOH
MW: 134.09 g/mol
Boiling Pt: 306 °C (1013 hPa)
Melting Pt: 131…132 °C
Density: 1.601 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064212
CAS Number: 617-48-1
EINECS: 210-514-9
Merck Index: 13,05730
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=920b54632a869bc5df22624fb9fecd4d11cae65e&vid=278193678126274041defdc0bd71c7c40efb3085&prid=web&clid=SAPDAM