(±)-sec-Butylamine, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
M-Clarity™ quality level = MQ100
B89000-100ML
B89000-500ML
SIALB89000-100MLEA
42.5
EUR
SIALB89000-100ML
SIALB89000-500ML
(±)-sec-Butylamine, Sigma-Aldrich®
(±)-sec-Butylamine
Formula:
C₄H₁₁N Boiling Pt: 63.5 °C (1013 hPa) Melting Pt: –104 °C Density: 0.724 g/cm³ (20 °C) Flash Pt: –20 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008094 CAS Number: 13952-84-6 EINECS: 237-732-7 UN: 2733 ADR: 3,II Merck Index: 12,01579 |