(±)-1-Aminopropan-2-ol, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Aminopropanol, Hydroxypropylamine, 1-Aminopropan-2-ol, DL-1-Amino-2-propanol, (±)-Isopropanolamine, Aminoisopropyl alcohol, iso-propanol amine Amino-2-propanol DL-Isopropanolamine (±)-1-Amino-2-propanol (±)-Isopropanolamine
MQ200
110248-100ML 110248-1L
SIAL110248-100MLEA 42.1 EUR
SIAL110248-100ML SIAL110248-1L
(±)-1-Aminopropan-2-ol, Sigma-Aldrich®
(±)-1-Aminopropan-2-ol
Formula: C₃H₉NO
MW: 75.11 g/mol
Boiling Pt: 158…160 °C (1013 hPa)
Melting Pt: 1
Density: 0.962 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008139
CAS Number: 78-96-6
EINECS: 201-162-7
UN: 2735
ADR: 8 III