(±)-2-Aminobutan-1-ol, Sigma-Aldrich®

Supplier: Merck
Danger

Synonyms: 2-Aminobutan-1-ol, (±)-2-Amino-1-butanol

M-Clarity™ quality level = MQ100

A43804-100ML A43804-500ML
SIALA43804-100MLEA 53.1 EUR
SIALA43804-100ML SIALA43804-500ML
(±)-2-Aminobutan-1-ol, Sigma-Aldrich®
(±)-2-Aminobutan-1-ol
Formula: C₄H₁₁NO
MW: 89.14 g/mol
Boiling Pt: 176 °C (1013 hPa)
Melting Pt: –2 °C
Density: 0.944 g/cm³ (20 °C)
Flash Pt: 91 °C
Storage Temperature: Ambient
MDL Number: MFCD00008095
CAS Number: 96-20-8
EINECS: 202-488-2
UN: 2735
ADR: 8,III
Merck Index: 13,00426

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR