(±)-1-Methoxy-2-propanol ≥99%
Supplier: Thermo Fisher Scientific
Danger
Synonyms: (±)-Monopropylene glycol methyl ether, 1-Methoxy-2-propanol
L12694.AK L12694.AU
L12694.AKEA 26.8 EUR
L12694.AK L12694.AU
(±)-1-Methoxy-2-propanol ≥99%
(±)-1-Methoxy-2-propanol
Formula: CH₃CH(OH)CH₂OCH₃
MW: 90.12 g/mol
Boiling Pt: 119.6
Melting Pt: –97 °C
Density: 0.92 g/cm³ (20 °C)
Flash Pt: 31
Storage Temperature: Ambient
MDL Number: MFCD00004537
CAS Number: 107-98-2
EINECS: 203-539-1
UN: 3092
ADR: 3 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f9e076d8e7a4ef95472e37d2c355f37d64eac8ee&vid=26da88ae9f09b8195abc9bdd949f6bbdaf429255&prid=web&clid=SAPDAM