(±)-1,2-Dodecanediol, Sigma-Aldrich®
Supplier: Merck
Synonyms:
(±)-1,2-Dihydroxy dodecane, 1,2-Dihydroxydodecane, 1,2-Dodecanediol
M-Clarity™ quality level = MQ100
213721-250G
213721-50G
SIAL213721-250GEA
370
EUR
SIAL213721-250G
SIAL213721-50G
(±)-1,2-Dodecanediol, Sigma-Aldrich®
(±)-1,2-Dodecanediol
Formula:
C₁₂H₂₆O₂ MW: 202.34 g/mol |
MDL Number:
MFCD00004726 CAS Number: 1119-87-5 EINECS: 214-289-8 |