(±)-1,3-Butanediol, Sigma-Aldrich®
Supplier: Merck
Synonyms: Butane-1,3-diol, (±)-1,3-Butylene glycol, (±)-1,3-Dihydroxy butane
M-Clarity™ quality level = MQ200
B84785-100ML B84785-1L B84785-20L B84785-2.5L
SIALB84785-100MLEA 42.9 EUR
SIALB84785-100ML SIALB84785-1L SIALB84785-20L SIALB84785-2.5L
(±)-1,3-Butanediol, Sigma-Aldrich®
(±)-1,3-Butanediol
Formula: C₄H₁₀O₂
MW: 90.12 g/mol
Boiling Pt: 206…208 °C (1013 hPa)
Melting Pt: < –50 °C
Density: 1.004…1.006 g/cm³ (20 °C)
Flash Pt: 116 °C
Storage Temperature: Ambient
MDL Number: MFCD00004554
CAS Number: 107-88-0
EINECS: 203-529-7
Merck Index: 13,01566