3-Mercapto-1,2-propanediol ≥97%, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: alpha-Thioglycerol, α-Monothioglycerol, Monothioglycerol, (±)-3-Mercapto-1,2-dihydroxy propane, 1-Thioglycerol, (±)-α-Monothioglycerine, (±)-1-Thioglycerine, (±)-3-Mercapto-1,2-propanediol
M-Clarity™ quality level = MQ100
M1753-100ML M1753-500ML
SIALM1753-100MLEA 200 EUR
SIALM1753-100ML SIALM1753-500ML
3-Mercapto-1,2-propanediol ≥97%, Sigma-Aldrich®
3-Mercapto-1,2-propanediol
Formula: C₃H₈O₂S
Boiling Pt: 118 °C/5 mmHg (lit.)
Density: 1.25 g/mL (25 °C) (lit.)
Storage Temperature: Refrigerator
MDL Number: MFCD00004879
CAS Number: 96-27-5
EINECS: 202-495-0
Merck Index: 12,09471