(±)-2-Methylbutyric acid, Sigma-Aldrich®
Supplier: Merck
Synonyms:
(±)-2-Methylbutanoic acid, 2-Methylbutyric acid, 2-Methylbutanoic acid
M-Clarity™ quality level = MQ100
W269514-100G-K
W269514-1KG-K
W269514-5KG-K
SIALW269514-100G-KEA
84.5
EUR
SIALW269514-100G-K
SIALW269514-1KG-K
SIALW269514-5KG-K
(±)-2-Methylbutyric acid, Sigma-Aldrich®
(±)-2-Methylbutyric acid
Formula:
C₅H₁₀O₂ Boiling Pt: 177 °C (1013 hPa) Melting Pt: –70 °C Density: 0.938 g/cm³ (20 °C) Flash Pt: 80 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002669 CAS Number: 116-53-0 EINECS: 204-145-2 UN: 3265 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...