(±)-2-Ethyl-1-hexanol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: 2-Ethylhexan-1-ol
MQ100
538051-1L 538051-4L 538051-500ML
SIAL538051-1LEA 46.5 EUR
SIAL538051-1L SIAL538051-4L SIAL538051-500ML
(±)-2-Ethyl-1-hexanol, Sigma-Aldrich®
(±)-2-Ethyl-1-hexanol
Formula: CH₃(CH₂)₃CH(C₂H₅)CH₂OH
MW: 130.23 g/mol
Boiling Pt: 183
Melting Pt: –76 °C
Density: 0.83 g/cm³ (20 °C)
Flash Pt: 81
Storage Temperature: Ambient
MDL Number: MFCD00004746
CAS Number: 104-76-7
EINECS: 203-234-3
REACH: 01-2119487289-20
Merck Index: 13,03842
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cc81f9275fbd3a7f8fb0e453419eab1652a4fdec&vid=1f7059eb6b27d30ac2c7e6b83f6bc0dff6002fe7&prid=web&clid=SAPDAM