(±)-2-Methyl-1-butanol, Sigma-Aldrich®
Supplier: Merck
|
Danger
|
M-Clarity™ quality level = MQ200
133051-100ML
133051-1L
SIAL133051-100MLEA
33.4
EUR
SIAL133051-100ML
SIAL133051-1L
(±)-2-Methyl-1-butanol, Sigma-Aldrich®
(±)-2-Methyl-1-butanol
Formula:
C₅H₁₂O MW: 88.15 g/mol Boiling Pt: 129 °C (1013 hPa) Melting Pt: –70 °C Density: 0.82 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00004743 CAS Number: 137-32-6 EINECS: 205-289-9 UN: 1105 ADR: 3,III Merck Index: 12,06109 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...