(±)-3-Methyl-2-butanol 98%
Supplier: Thermo Fisher Scientific
Warning
Synonyms: DL-3-Methyl-2-butanol
149930250 149931000 149935000
ACRO149930250EA 24.7 EUR
ACRO149930250 ACRO149931000 ACRO149935000
(±)-3-Methyl-2-butanol 98%
(±)-3-Methyl-2-butanol
Formula: C₅H₁₂O
MW: 88.15 g/mol
Boiling Pt: 112 °C (760 mmHg)
Melting Pt: -117
Density: 0.81 g/cm³
Flash Pt: 39 °C
Storage Temperature: Ambient
MDL Number: MFCD00004527
CAS Number: 598-75-4
EINECS: 209-950-2
UN: 1105
ADR: 3 III
Merck Index: 12,06110
Order Now
Specification Test Results
Appearance Clear colorless Liquid
Infrared spectrum Conforms
Refractive index 1.4080 to 1.4100 (20°C, 589 nm)
GC ≥97.5 %