(±)-3-Methyl-2-butanol, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: DL-3-Methyl-2-butanol
M-Clarity™ quality level = MQ100
110949-100ML 110949-500ML
SIAL110949-100MLEA 77.8 EUR
SIAL110949-100ML SIAL110949-500ML
(±)-3-Methyl-2-butanol, Sigma-Aldrich®
(±)-3-Methyl-2-butanol
Formula: C₅H₁₂O
MW: 88.15 g/mol
Boiling Pt: 112 °C (1013 hPa)
Melting Pt: –117 °C
Density: 0.82 g/cm³ (20 °C)
Flash Pt: 26 °C
Storage Temperature: Ambient
MDL Number: MFCD00004527
CAS Number: 598-75-4
EINECS: 209-950-2
UN: 1105
ADR: 3,III
Merck Index: 12,06110