(±)-4-Methyl-2-pentanol, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: (±)-Methyl isobutyl carbinol
M-Clarity™ quality level = MQ200
109916-100ML 109916-1L
SIAL109916-100MLEA 30.3 EUR
SIAL109916-100ML SIAL109916-1L
(±)-4-Methyl-2-pentanol, Sigma-Aldrich®
(±)-4-Methyl-2-pentanol
Formula: C₆H₁₄O
MW: 102.18 g/mol
Boiling Pt: 132 °C (1013 hPa)
Melting Pt: –90 °C
Density: 0.8093 g/cm³ (20 °C)
Flash Pt: 54 °C
Storage Temperature: Ambient
MDL Number: MFCD00004550
CAS Number: 108-11-2
EINECS: 203-551-7
UN: 2053
ADR: 3,III