DL-Tryptophan, Sigma-Aldrich®

Supplier: Merck

Synonyms: H-DL-Trp-OH, (+/-)-2-Amino-3-(3-indolyl)propionic acid, 2-Amino-3-(1H-indol-3-yl)propanoic acid

M-Clarity™ quality level = MQ200

162698-50G 162698-1.5G
SIAL162698-50GEA 153 EUR
SIAL162698-50G SIAL162698-1.5G
DL-Tryptophan, Sigma-Aldrich®
DL-Tryptophan
Formula: C₁₁H₁₂N₂O₂
MW: 204.23 g/mol
Melting Pt: 289…290 °C
Density: 1.349 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064339
CAS Number: 54-12-6
EINECS: 200-194-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR