5-Fluoro-DL-tryptophan
Supplier: Thermo Scientific
Synonyms: 2-Amino-3-(5-fluoro-1H-indol-3-yl)propanoic acid, H-DL-Trp(5-F)-OH
AC11968.250 AC11968.1
MAYBAC11968.250EA 0 EUR
MAYBAC11968.250 MAYBAC11968.1
5-Fluoro-DL-tryptophan
5-Fluoro-DL-tryptophan
Formula: C₁₁H₁₁FN₂O₂
MW: 222.22 g/mol
Melting Pt: 263…264 °C
Storage Temperature: Ambient
MDL Number: MFCD00005649
CAS Number: 154-08-5
EINECS: 205-822-5
Order Now
Specification Test Results
Appearance Light beige Powder