DL-m-Tyrosine 98.5%
Supplier: Thermo Fisher Scientific
Warning
140630010 140630050
ACRO140630010EA 112 EUR
ACRO140630010 ACRO140630050
DL-m-Tyrosine 98.5%
DL-m-Tyrosine
Formula: C₉H₁₁NO₃
Boiling Pt: 351 °C (1013 hPa)
Melting Pt: 280 °C (dec.)
Density: 1,456 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002597
CAS Number: 775-06-4
EINECS: 212-270-9
Merck Index: 12,09971
Order Now
Specification Test Results
Appearance White to light beige Crystalline powder
Infrared spectrum Conforms
Water ≤0.5 % (K.F.)
Titration with HClO4 ≥98.0 %