(±)-1-Hepten-3-ol, Sigma-Aldrich®
Supplier: Merck
Synonyms: 1-Hepten-3-ol, (±)-Hept-1-en-3-ol
M-Clarity™ quality level = MQ300
W412901-100G W412901-25G W412901-SAMPLE
SIALW412901-100GEA 405 EUR
SIALW412901-100G SIALW412901-25G SIALW412901-SAMPLE
(±)-1-Hepten-3-ol, Sigma-Aldrich®
(±)-1-Hepten-3-ol
Formula: C₇H₁₄O
MW: 114.19 g/mol
Boiling Pt: 154 °C (1013 hPa)
Density: 0.8396 g/cm³ (20 °C)
MDL Number: MFCD00021940
CAS Number: 4938-52-7
EINECS: 225-579-9
UN: 1987
ADR: 3,III