(±)-1-Hexen-3-ol, Sigma-Aldrich®
Supplier: Merck
Synonyms: hex-1-En-3-ol, (±)-Hex-1-en-3-ol
M-Clarity™ quality level = MQ300
W360805-1KG-K W360805-25G-K
SIALW360805-1KG-KEA 615 EUR
SIALW360805-1KG-K SIALW360805-25G-K
(±)-1-Hexen-3-ol, Sigma-Aldrich®
(±)-1-Hexen-3-ol
Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 134…135 °C (1013 hPa)
Melting Pt: –72 °C
Density: 0.884 g/cm³ (20 °C)
MDL Number: MFCD00004581
CAS Number: 4798-44-1
EINECS: 225-355-0
UN: 1987
ADR: 3,III