(±)-1-Hepten-4-ol ≥97%
Supplier: Thermo Scientific
Warning
B20591.03 B20591.06 B20591.14
B20591.03EA 32.5 EUR
B20591.03 B20591.06 B20591.14
(±)-1-Hepten-4-ol ≥97%
(±)-1-Hepten-4-ol
Purity: 97% CAS No.: 3521-91-3 Molecular Formula: C7H14O Formula Weight: 114.18 UN No.: 1987 MDL No.: MFCD00021933
Formula: C₇H₁₄O
MW: 114.19 g/mol
Boiling Pt: 152.1 °C (1013 hPa)
Melting Pt: –61 °C
Density: 0.8412 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00021933
CAS Number: 3521-91-3
EINECS: 222-535-0
UN: 1987
ADR: 3,III