(±)-3-Butyne-2-ol, Sigma-Aldrich®
Supplier: Merck
Synonyms: (±)-But-3-yn-2-ol, (±)-3-Hydroxy-1-butyne
M-Clarity™ quality level = MQ100
447986-100ML 447986-25ML
SIAL447986-100MLEA 262 EUR
SIAL447986-100ML SIAL447986-25ML
(±)-3-Butyne-2-ol, Sigma-Aldrich®
(±)-3-Butyne-2-ol
Formula: C₄H₆O
MW: 70.09 g/mol
Boiling Pt: 104…108 °C (1013 hPa)
Melting Pt: –12 °C
Density: 0.895 g/cm³ (20 °C)
MDL Number: MFCD00004541
CAS Number: 2028-63-9
EINECS: 217-978-1
UN: 2929
ADR: 6.1,II