(±)-1-Heptyn-3-ol, Sigma-Aldrich®

Supplier: Merck

Synonyms: Hept-1-yn-3-ol

M-Clarity™ quality level = MQ100

666963-10G 666963-1G
SIAL666963-10GEA 165 EUR
SIAL666963-10G SIAL666963-1G
(±)-1-Heptyn-3-ol, Sigma-Aldrich®
(±)-1-Heptyn-3-ol
Formula: C₇H₁₂O
MW: 112.17 g/mol
Boiling Pt: 161.5…162.5 °C (1013 hPa)
Density: 0.87 g/cm³ (20 °C)
Storage Temperature: Refrigerator
MDL Number: MFCD00041597
CAS Number: 7383-19-9
UN: 1987
ADR: 3,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR