(±)-1-Octyn-3-ol, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: Oct-1-yn-3-ol
M-Clarity™ quality level = MQ100
127280-250G 127280-50G
SIAL127280-250GEA 363 EUR
SIAL127280-250G SIAL127280-50G
(±)-1-Octyn-3-ol, Sigma-Aldrich®
(±)-1-Octyn-3-ol
Formula: C₈H₁₄O
MW: 126.2 g/mol
Boiling Pt: 182
Melting Pt: 0
Density: 0.864 g/cm³ (20 °C)
MDL Number: MFCD00004588
CAS Number: 818-72-4
EINECS: 212-455-4
UN: 2810
ADR: 6.1 III