1-Phenyl-2-propanol ≥98%, Sigma-Aldrich®
Supplier: Merck
Synonyms: (2-Hydroxypropyl)benzol, (±)-1-Phenyl-2-propanol, (±)-α-Methylphenethyl alcohol, Benzylmethylcarbinol
M-Clarity™ quality level = MQ100
189235-100G 189235-25G 189235-5G
SIAL189235-100GEA 885 EUR
SIAL189235-100G SIAL189235-25G SIAL189235-5G
1-Phenyl-2-propanol ≥98%, Sigma-Aldrich®
1-Phenyl-2-propanol
Formula: C₆H₅CH₂CH(OH)CH₃
MW: 136.19 g/mol
Boiling Pt: 219…221 °C (1013 hPa)
Melting Pt: 65…67 °C
Density: 0.997 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004540
CAS Number: 698-87-3
EINECS: 211-821-0