DL-Camphor, Sigma-Aldrich®
Supplier: Merck
Warning
Synonyms: (±)-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone, Camphor, 1,7,7-Trimethylbicyclo[2.2.1]heptan-2-one, DL-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanone, (±)-1,7,7-Trimethyl-2-norbornanone
M-Clarity™ quality level = MQ100
148075-100G 148075-500G
SIAL148075-100GEA 34.6 EUR
SIAL148075-100G SIAL148075-500G
DL-Camphor, Sigma-Aldrich®
DL-Camphor
Formula: C₁₀H₁₆O
MW: 152.24 g/mol
Boiling Pt: 204 °C (1013 hPa)
Melting Pt: ∼180 °C
Density: 0.992 g/cm³ (25 °C)
Flash Pt: 66 °C
Storage Temperature: Ambient
MDL Number: MFCD00074738
CAS Number: 76-22-2
EINECS: 200-945-0
UN: 2717
ADR: 4.1,III