DL-Camphor, Sigma-Aldrich®
Supplier: Merck
M-Clarity™ quality level = MQ300
SIALW526606-25KG-KEA 1580 EUR
SIALW526606-25KG-K SIALW526606-1KG-K
DL-Camphor, Sigma-Aldrich®
DL-Camphor
Formula: C₁₀H₁₆O
MW: 152,24 g/mol
Boiling Pt: 204 °C (1013 hPa)
Melting Pt: ∼180 °C
Density: 0,992 g/cm³ (25 °C)
Flash Pt: 66 °C
MDL Number: MFCD00074738
UN: 2717
ADR: 4.1,III