α-Pinene, Sigma-Aldrich®

Supplier: Merck


M-Clarity™ quality level = MQ100

147524-1L 147524-250ML 147524-5ML
SIAL147524-1LEA 350 EUR
SIAL147524-1L SIAL147524-250ML SIAL147524-5ML
α-Pinene, Sigma-Aldrich®
α-Pinene
Formula: C₁₀H₁₆
MW: 136.24 g/mol
Boiling Pt: 155 °C (1013 hPa)
Melting Pt: –55 °C
Density: 0.8592 g/cm³ (20 °C)
MDL Number: MFCD00001339
CAS Number: 80-56-8
EINECS: 201-291-9
UN: 2368
ADR: 3,III
Merck Index: 12,07599

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR