(±)-trans-1,2-Cyclohexanediol, Sigma-Aldrich®
Supplier: Merck
Synonyms:
(±)-trans-Cyclohexane-1,2-diol, trans-Cyclohexane-1,2-diol
M-Clarity™ quality level = MQ100
141712-10G
141712-50G
SIAL141712-10GEA
48.2
EUR
SIAL141712-10G
SIAL141712-50G
(±)-trans-1,2-Cyclohexanediol, Sigma-Aldrich®
(±)-trans-1,2-Cyclohexanediol
Formula:
C₆H₁₂O₂ MW: 116.16 g/mol Boiling Pt: 237 °C (1013 hPa) Melting Pt: 100…103 °C Density: 1.147 g/cm³ (24 °C) |
MDL Number:
MFCD00063611 CAS Number: 1460-57-7 EINECS: 215-956-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...