Glutaraldehyde 25% in aqueous solution
Supplier: Roth Carl
Danger
Synonyms: Glutaral, Glutardialdehyde, 1,5-Pentanedial
3778.1 3778.2
ROTH3778.1EA 91 EUR
ROTH3778.1 ROTH3778.2
Glutaraldehyde 25% in aqueous solution
Glutaraldehyde
Formula: C₅H₈O₂
MW: 100.12 g/mol
Boiling Pt: 101 °C (1013 hPa)
Melting Pt: –6 °C
Density: 1.06 g/cm³ (20 °C)
Flash Pt: >71 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00007025
CAS Number: 111-30-8
EINECS: 203-856-5
UN: 2927
ADR: 6.1,II