Glutaraldehyde ≥70% in aqueous solution, Sigma-Aldrich®
Supplier: Merck
Danger
Synonyms: Glutaral, Glutardialdehyde, 1,5-Pentanedial
M-Clarity™ quality level = MQ200
G7776-10ML G7776-10X10ML
SIALG7776-10MLEA 120 EUR
SIALG7776-10ML SIALG7776-10X10ML
Glutaraldehyde ≥70% in aqueous solution, Sigma-Aldrich®
Glutaraldehyde
Formula: C₅H₈O₂
MW: 100.12 g/mol
MDL Number: MFCD00007025
CAS Number: 111-30-8
EINECS: 203-856-5
UN: 2927
ADR: 6.1,II