Hydrochloric acid 0.0714 mol/l (N/14)
Supplier: PROLABO CMP
MC307902.6010EA 0 EUR
MC307902.6010 PRLSMC307902.9010 PRLSMC307902.9020
Hydrochloric acid 0.0714 mol/l (N/14)
Hydrochloric acid
Formula: HCl
MW: 36,46 g/mol
Density: 1,004 g/cm³ (25 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,III