Hydrochloric acid 3 mol/l (3 N), VOLUSOL® volumetric solution
Supplier: VWR Chemicals
Warning
Synonyms: Hydrochloric acid solution, Muriatic acid
5050.5000EA 147 EUR
5050.5000 5050.1000
Hydrochloric acid 3 mol/l (3 N), VOLUSOL® volumetric solution
Hydrochloric acid
Formula: HCl
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 0
UN: 1789
ADR: 8,III
Order Now
Specification Test Results
Titer 2.95 - 3.05 mol/l