5-Aminopentan-1-ol 50% in aqueous solution
Supplier: Thermo Fisher Scientific
Danger
Synonyms: 5-Amino-1-pentanol
387911000 387918000 387910250
ACRO387911000EA 111 EUR
ACRO387911000 ACRO387918000 ACRO387910250
5-Aminopentan-1-ol 50% in aqueous solution
5-Aminopentan-1-ol
Formula: C₅H₁₃NO
MW: 103.16 g/mol
Boiling Pt: 222
Melting Pt: 36
Density: 0.987 g/cm³
Flash Pt: >100 °C
Storage Temperature: Ambient
MDL Number: MFCD00008237
CAS Number: 2508-29-4
EINECS: 219-718-2
UN: 2735
ADR: 8 III
Order Now
Specification Test Results
Appearance Clear colorless Solution
Infrared spectrum Conforms
Titration with HCl 48.0 to 52.0 %